Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-hepten-4-one, predominantly trans, 99%

Click to view available options
Quantity:
2g
10g
81925-81-7
C8H14O
126.199
MFCD00792509
ARJWAURHQDJJAC-GQCTYLIASA-N
5-methyl-2-hepten-4-one,filbertone,5-methylhept-2-en-4-one,+/--filbertone,hazelnut ketone,e-5-methyl-2-hepten-4-one,5-methyl-e-2-hepten-4-one,fema no. 3761,e-5-methylhept-2-en-4-one,2e-5-methyl-2-hepten-4-one
5362588
(E)-5-methylhept-2-en-4-one
CCC(C)C(=O)C=CC
81925-81-7
C8H14O
126.199
MFCD00792509
ARJWAURHQDJJAC-GQCTYLIASA-N
5-methyl-2-hepten-4-one,filbertone,5-methylhept-2-en-4-one,+/--filbertone,hazelnut ketone,e-5-methyl-2-hepten-4-one,5-methyl-e-2-hepten-4-one,fema no. 3761,e-5-methylhept-2-en-4-one,2e-5-methyl-2-hepten-4-one
5362588
(E)-5-methylhept-2-en-4-one
CCC(C)C(=O)C=CC
Flash Point 55°C (131°F)
Boiling Point 169°C to 171°C
Quantity 2g
UN Number UN1224
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hepten-4-one, predominantly trans

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : TSCA