Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-hexanol, 98%

Click to view available options
Quantity:
5g
25g
627-59-8
C7H16O
116.204
MFCD00004558
ZDVJGWXFXGJSIU-UHFFFAOYSA-N
5-methyl-2-hexanol,2-hexanol, 5-methyl,2-methyl-5-hexanol,2-hexanol,5-methyl,5-methylhexanol-2,acmc-1b03m,zdvjgwxfxgjsiu-uhfffaoysa
12323
5-methylhexan-2-ol
CC(C)CCC(C)O
627-59-8
C7H16O
116.204
MFCD00004558
ZDVJGWXFXGJSIU-UHFFFAOYSA-N
5-methyl-2-hexanol,2-hexanol, 5-methyl,2-methyl-5-hexanol,2-hexanol,5-methyl,5-methylhexanol-2,acmc-1b03m,zdvjgwxfxgjsiu-uhfffaoysa
12323
5-methylhexan-2-ol
CC(C)CCC(C)O
Quantity 25g
Boiling Point 148°C to 150°C
Flash Point 46°C (114°F)
UN Number UN1987
Beilstein 1731619
Formula Weight 116.2
Percent Purity 98%
Chemical Name or Material 5-Methyl-2-hexanol

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.