Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-hexanol, 98%, Acros Organics

Click to view available options
Quantity:
10g
627-59-8
C7H16O
116.204
MFCD00004558
ZDVJGWXFXGJSIU-UHFFFAOYSA-N
5-methyl-2-hexanol,2-hexanol, 5-methyl,2-methyl-5-hexanol,2-hexanol,5-methyl,5-methylhexanol-2,acmc-1b03m,5-methyl-2-hexanol,zdvjgwxfxgjsiu-uhfffaoysa
12323
5-methylhexan-2-ol
CC(C)CCC(C)O
627-59-8
C7H16O
116.204
MFCD00004558
ZDVJGWXFXGJSIU-UHFFFAOYSA-N
5-methyl-2-hexanol,2-hexanol, 5-methyl,2-methyl-5-hexanol,2-hexanol,5-methyl,5-methylhexanol-2,acmc-1b03m,5-methyl-2-hexanol,zdvjgwxfxgjsiu-uhfffaoysa
12323
5-methylhexan-2-ol
CC(C)CCC(C)O
Quantity 10g
Density 0.81g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2CHCH2CH2CH(OH)CH3
Beilstein 01, 416
Formula Weight 116.2
Chemical Name or Material 5-Methyl-2-hexanol, 98%

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning