Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-nitrobenzoic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3113-72-2
C8H7NO4
181.15
QRRSIFNWHCKMSW-UHFFFAOYSA-N
5-methyl-2-nitrobenzoic acid,2-nitro-5-methylbenzoic acid,benzoic acid, 5-methyl-2-nitro,3-methyl-6-nitrobenzoic acid,6-nitro-m-toluic acid,m-toluic acid, 6-nitro,6-nitro-p-toluic acid,5-methyl-2-nitrobenzoicacid,5-methyl-2-nitro-benzoic acid,benzoic acid,5-methyl-2-nitro
18371
CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O
3113-72-2
C8H7NO4
181.15
QRRSIFNWHCKMSW-UHFFFAOYSA-N
5-methyl-2-nitrobenzoic acid,2-nitro-5-methylbenzoic acid,benzoic acid, 5-methyl-2-nitro,3-methyl-6-nitrobenzoic acid,6-nitro-m-toluic acid,m-toluic acid, 6-nitro,6-nitro-p-toluic acid,5-methyl-2-nitrobenzoicacid,5-methyl-2-nitro-benzoic acid,benzoic acid,5-methyl-2-nitro
18371
CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O
Packaging Glass bottle
Color White to Yellow
Quantity 25g
Melting Point 133.0°C to 136.0°C
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3C6H3(NO2)CO2H
Beilstein 09,482
Formula Weight 181.15
Physical Form Powder or Crystalline Powder
Percent Purity 95%
Chemical Name or Material 5-Methyl-2-nitrobenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.