Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-octanone, tech. 75%, cont. 5-nonanone ca 10%

Manufacturer:  Alfa Aesar™ B2516009

Quantity 10g

 View more versions of this product

Catalog No. AAB2516009


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

58654-67-4
142.24
5-Methyl-2-octanone
C9H18O
MFCD00059414
Technical Grade

Specifications

58654-67-4
142.24
10g
75%, cont. 5-nonanone ≈10%
C9H18O
MFCD00059414
5-Methyl-2-octanone
Technical Grade

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit