Alfa Aesar™ 5-Methyl-2-octanone, tech. 75%, cont. 5-nonanone ca 10%

Manufacturer: Alfa Aesar™ B2516030

Quantity 250g

 View more versions of this product

Catalog No. AAB2516030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 58654-67-4
    Molecular Formula C9H18O
    Formula Weight 142.24
    MDL Number MFCD00059414
    Chemical Name or Material 5-Methyl-2-octanone
    Grade Technical Grade

    Specifications

    Formula Weight 142.24
    TSCA TSCA
    Molecular Formula C9H18O
    MDL Number MFCD00059414
    CAS 58654-67-4
    Quantity 250g
    Chemical Name or Material 5-Methyl-2-octanone
    Assay 75%, cont. 5-nonanone ≈10%
    Grade Technical Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit