Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-octanone, tech. 75%, cont. 5-nonanone ca 10%

Click to view available options
Quantity:
10g
50g
250g
58654-67-4
C9H18O
142.242
MFCD00059414
UPVLGOPSYZTLGB-UHFFFAOYSA-N
5-methyl-2-octanone,2-octanone, 5-methyl,methyl 3-methylhexyl ketone,methyloctanone,acmc-1axv1,dsstox_cid_21444,dsstox_gsid_41444,3-01-00-02890 beilstein handbook reference
42802
5-methyloctan-2-one
CCCC(C)CCC(=O)C
58654-67-4
C9H18O
142.242
MFCD00059414
UPVLGOPSYZTLGB-UHFFFAOYSA-N
5-methyl-2-octanone,2-octanone, 5-methyl,methyl 3-methylhexyl ketone,methyloctanone,acmc-1axv1,dsstox_cid_21444,dsstox_gsid_41444,3-01-00-02890 beilstein handbook reference
42802
5-methyloctan-2-one
CCCC(C)CCC(=O)C
Quantity 50g
Formula Weight 142.24
Percent Purity 75%
Grade Technical
Chemical Name or Material 5-Methyl-2-octanone
Assay cont. 5-nonanone ≈10%