Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-thiophenecarbaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
13679-70-4
C6H6OS
126.18
MFCD00005434
VAUMDUIUEPIGHM-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxaldehyde,5-methylthiophene-2-carboxaldehyde,5-methyl-2-thiophenecarbaldehyde,2-formyl-5-methylthiophene,5-methyl-2-formylthiophene,2-thiophenecarboxaldehyde, 5-methyl,5-methylthiophene-2-aldehyde,5-methyl-2-thenaldehyde,5-methyl-2-thiophencarboxaldehyde,5-methyl-2-thiophene carboxaldehyde
61663
5-methylthiophene-2-carbaldehyde
CC1=CC=C(S1)C=O
13679-70-4
C6H6OS
126.18
MFCD00005434
VAUMDUIUEPIGHM-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxaldehyde,5-methylthiophene-2-carboxaldehyde,5-methyl-2-thiophenecarbaldehyde,2-formyl-5-methylthiophene,5-methyl-2-formylthiophene,2-thiophenecarboxaldehyde, 5-methyl,5-methylthiophene-2-aldehyde,5-methyl-2-thenaldehyde,5-methyl-2-thiophencarboxaldehyde,5-methyl-2-thiophene carboxaldehyde
61663
5-methylthiophene-2-carbaldehyde
CC1=CC=C(S1)C=O
Specific Gravity 1.18
Flash Point 87°C
Refractive Index 1.5800 to 1.584
Color Yellow to Brown
Quantity 10mL
Infrared Spectrum Authentic
Density 1.1800g/mL
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 17, I, 151
Formula Weight 126.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 5-Methyl-2-thiophenecarboxaldehyde
Show More Show Less