Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-thiophenecarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
Packaging:
Glass bottle
1918-79-2
C6H6O2S
142.18
MFCD00005439
VCNGNQLPFHVODE-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxylic acid,5-methyl-2-thenoic acid,2-thiophenecarboxylic acid, 5-methyl,2-carboxy-5-methylthiophene,5-methly-2-thiophenecarboxylic acid,5-methyl-thiophene-2-carboxylic acid,5-methyl-2-thiophene carboxylic acid,5-methylthiophen-2-carboxylic acid,2-methyl-5-thiophenecarboxylic acid,pubchem5206
74713
5-methylthiophene-2-carboxylic acid
CC1=CC=C(S1)C(=O)O
1918-79-2
C6H6O2S
142.18
MFCD00005439
VCNGNQLPFHVODE-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxylic acid,5-methyl-2-thenoic acid,2-thiophenecarboxylic acid, 5-methyl,2-carboxy-5-methylthiophene,5-methly-2-thiophenecarboxylic acid,5-methyl-thiophene-2-carboxylic acid,5-methyl-2-thiophene carboxylic acid,5-methylthiophen-2-carboxylic acid,2-methyl-5-thiophenecarboxylic acid,pubchem5206
74713
5-methylthiophene-2-carboxylic acid
CC1=CC=C(S1)C(=O)O
Quantity 2.5g
Melting Point 136°C to 138°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 18, 295
Formula Weight 142.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-thiophenecarboxylic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 217-640-3