Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-thiophenecarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
Packaging:
Glass Bottle
1918-79-2
C6H6O2S
142.172
MFCD00005439
VCNGNQLPFHVODE-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxylic acid,5-methyl-2-thenoic acid,2-thiophenecarboxylic acid, 5-methyl,2-carboxy-5-methylthiophene,5-methly-2-thiophenecarboxylic acid,5-methyl-thiophene-2-carboxylic acid,5-methyl-2-thiophene carboxylic acid,5-methylthiophen-2-carboxylic acid,2-methyl-5-thiophenecarboxylic acid,pubchem5206
74713
5-methylthiophene-2-carboxylic acid
CC1=CC=C(S1)C(=O)O
1918-79-2
C6H6O2S
142.172
MFCD00005439
VCNGNQLPFHVODE-UHFFFAOYSA-N
5-methyl-2-thiophenecarboxylic acid,5-methyl-2-thenoic acid,2-thiophenecarboxylic acid, 5-methyl,2-carboxy-5-methylthiophene,5-methly-2-thiophenecarboxylic acid,5-methyl-thiophene-2-carboxylic acid,5-methyl-2-thiophene carboxylic acid,5-methylthiophen-2-carboxylic acid,2-methyl-5-thiophenecarboxylic acid,pubchem5206
74713
5-methylthiophene-2-carboxylic acid
CC1=CC=C(S1)C(=O)O
Infrared Spectrum Authentic
Melting Point 136°C to 139°C
Quantity 10g
Packaging Glass Bottle
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 18, 295
Formula Weight 142.18
Physical Form Liquid
Percent Purity 98.5 to 101.5% (Acid)
Chemical Name or Material 5-Methyl-2-thiophenecarboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.