Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-3-heptyl Acetate 95.0+%, TCI America™

Click to view available options
Quantity:
5g
375855-08-6
C10H20O2
172.268
MFCD00671490
KCEXFYWUSJAMSL-UHFFFAOYSA-N
Acetic Acid 1-Ethyl-3-methylpentyl Ester, Acetic Acid 5-Methyl-3-heptyl Ester, 1-Ethyl-3-methylpentyl Acetate
44630060
5-methylheptan-3-yl acetate
CCC(C)CC(CC)OC(=O)C
375855-08-6
C10H20O2
172.268
MFCD00671490
KCEXFYWUSJAMSL-UHFFFAOYSA-N
Acetic Acid 1-Ethyl-3-methylpentyl Ester, Acetic Acid 5-Methyl-3-heptyl Ester, 1-Ethyl-3-methylpentyl Acetate
44630060
5-methylheptan-3-yl acetate
CCC(C)CC(CC)OC(=O)C
Boiling Point 185°C
Quantity 5g
Color Colorless
Formula Weight 172.27
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material 5-Methyl-3-heptyl Acetate

EINECSNumber : (2)-0735

TSCA : No