Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-3-trifluoromethyl-1H-pyrazole, 98%

Click to view available options
Quantity:
250mg
1g
5g
10010-93-2
C5H5F3N2
150.104
MFCD00156058
DLCHCAYDSKIFIN-UHFFFAOYSA-N
5-methyl-3-trifluoromethyl-1h-pyrazole,3-methyl-5-trifluoromethyl-1h-pyrazole,3-methyl-5-trifluoromethyl pyrazole,1h-pyrazole, 3-methyl-5-trifluoromethyl,5-methyl-3-trifluoromethyl pyrazole,3-trifluoromethyl-5-methyl pyrazole,pyrazole, 3-methyl-5-trifluoromethyl,3-methyl-5-trifluoromethylpyrazole,3-trifluoromethyl-5-methylpyrazole,3-methyl-5-trifluoromethyl-2h-pyrazole
139077
5-methyl-3-(trifluoromethyl)-1H-pyrazole
CC1=CC(=NN1)C(F)(F)F
10010-93-2
C5H5F3N2
150.104
MFCD00156058
DLCHCAYDSKIFIN-UHFFFAOYSA-N
5-methyl-3-trifluoromethyl-1h-pyrazole,3-methyl-5-trifluoromethyl-1h-pyrazole,3-methyl-5-trifluoromethyl pyrazole,1h-pyrazole, 3-methyl-5-trifluoromethyl,5-methyl-3-trifluoromethyl pyrazole,3-trifluoromethyl-5-methyl pyrazole,pyrazole, 3-methyl-5-trifluoromethyl,3-methyl-5-trifluoromethylpyrazole,3-trifluoromethyl-5-methylpyrazole,3-methyl-5-trifluoromethyl-2h-pyrazole
139077
5-methyl-3-(trifluoromethyl)-1H-pyrazole
CC1=CC(=NN1)C(F)(F)F
Color White
Melting Point 88°C to 90°C
Quantity 5g
Beilstein 743044
Formula Weight 150.1
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 5-Methyl-3-trifluoromethyl-1H-pyrazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning