Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-5-nitroimidazole, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
14003-66-8
C4H5N3O2
127.1
WSYOWIMKNNMEMZ-UHFFFAOYSA-N
4-methyl-5-nitroimidazole,5-methyl-4-nitroimidazole,4-methyl-5-nitro-1h-imidazole,1h-imidazole, 4-methyl-5-nitro,imidazole, 4-methyl-5-nitro,imidazole, 5-methyl-4-nitro,unii-5pul8bw840,imidazole, 4 or 5-methyl-5 or 4-nitro,4-methyl-5-nitro-3h-imidazole,1h-imidazole,4-methyl-5-nitro
26385
5-methyl-4-nitro-1H-imidazole
CC1=C(N=CN1)[N+](=O)[O-]
14003-66-8
C4H5N3O2
127.1
WSYOWIMKNNMEMZ-UHFFFAOYSA-N
4-methyl-5-nitroimidazole,5-methyl-4-nitroimidazole,4-methyl-5-nitro-1h-imidazole,1h-imidazole, 4-methyl-5-nitro,imidazole, 4-methyl-5-nitro,imidazole, 5-methyl-4-nitro,unii-5pul8bw840,imidazole, 4 or 5-methyl-5 or 4-nitro,4-methyl-5-nitro-3h-imidazole,1h-imidazole,4-methyl-5-nitro
26385
5-methyl-4-nitro-1H-imidazole
CC1=C(N=CN1)[N+](=O)[O-]
Melting Point 203.2°C to 203.3°C
Packaging Glass bottle
Color White to Brown
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 127.1
Physical Form Powder or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Methyl-5-nitroimidazole
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-809-5