Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-5-phenylhydantoin, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6843-49-8
C10H10N2O2
190.2
MFCD00005265
JNGWGQUYLVSFND-UHFFFAOYSA-N
5-methyl-5-phenylhydantoin,hydantoin, 5-methyl-5-phenyl,2,4-imidazolidinedione,5-methyl-5-phenyl,2,4-imidazolidinedione, 5-methyl-5-phenyl,mpid standard,acmc-20m4pt,2,4-imidazolidinedione, 5-methyl-5-phenyl-, r,2, 5-methyl-5-phenyl,+/--5-methyl-5-phenylhydantoin
93043
5-methyl-5-phenylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1)C2=CC=CC=C2
6843-49-8
C10H10N2O2
190.2
MFCD00005265
JNGWGQUYLVSFND-UHFFFAOYSA-N
5-methyl-5-phenylhydantoin,hydantoin, 5-methyl-5-phenyl,2,4-imidazolidinedione,5-methyl-5-phenyl,2,4-imidazolidinedione, 5-methyl-5-phenyl,mpid standard,acmc-20m4pt,2,4-imidazolidinedione, 5-methyl-5-phenyl-, r,2, 5-methyl-5-phenyl,+/--5-methyl-5-phenylhydantoin
93043
5-methyl-5-phenylimidazolidine-2,4-dione
CC1(C(=O)NC(=O)N1)C2=CC=CC=C2
Quantity 5g
Melting Point 199°C to 201°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 24, I, 347
Formula Weight 190.2
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 5-Methyl-5-phenylhydantoin
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Warning

EINECSNumber : 229-928-6