Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-DL-tryptophan, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
951-55-3
C12H14N2O2
218.25
HUNCSWANZMJLPM-UHFFFAOYSA-N
5-methyl-dl-tryptophan,5-methyltryptophan,2-amino-3-5-methyl-1h-indol-3-yl propanoic acid,tryptophan, 5-methyl,dl-5-methyltryptophan,5-methyl-tryptophan,2-amino-3-5-methylindol-3-yl propanoic acid,5-methyl tryptophan, dl,5-methyltryptophan #,pubchem20652
92852
CHEBI:52524
2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
CC1=CC2=C(C=C1)NC=C2CC(C(=O)O)N
951-55-3
C12H14N2O2
218.25
HUNCSWANZMJLPM-UHFFFAOYSA-N
5-methyl-dl-tryptophan,5-methyltryptophan,2-amino-3-5-methyl-1h-indol-3-yl propanoic acid,tryptophan, 5-methyl,dl-5-methyltryptophan,5-methyl-tryptophan,2-amino-3-5-methylindol-3-yl propanoic acid,5-methyl tryptophan, dl,5-methyltryptophan #,pubchem20652
92852
CHEBI:52524
2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
CC1=CC2=C(C=C1)NC=C2CC(C(=O)O)N
Quantity 5g
Appearance (5% soln. in 1N NaOH) clear, pale yellowish
Sulfated Ash 0.5% max.
Melting Point 280.0°C to 282.0°C
Water 1% Max. (K.F.)
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 22,II,471
Formula Weight 218.25
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 5-Methyl-DL-tryptophan
Show More Show Less