Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylbenzo[b]thiophene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
14315-14-1
C9H8S
148.23
MFCD00052509
DOHZWDWNQFZIKH-UHFFFAOYSA-N
5-methylbenzo b thiophene,benzo b thiophene, 5-methyl,5-methylbenzothiophene,5-methyl-benzo b thiophene,5-methylthianaphthene,5-methyl-benzothiophene,benzothiophene, 5-mehyl,5-methyl benzo b thiophene
84346
5-methyl-1-benzothiophene
CC1=CC2=C(C=C1)SC=C2
14315-14-1
C9H8S
148.23
MFCD00052509
DOHZWDWNQFZIKH-UHFFFAOYSA-N
5-methylbenzo b thiophene,benzo b thiophene, 5-methyl,5-methylbenzothiophene,5-methyl-benzo b thiophene,5-methylthianaphthene,5-methyl-benzothiophene,benzothiophene, 5-mehyl,5-methyl benzo b thiophene
84346
5-methyl-1-benzothiophene
CC1=CC2=C(C=C1)SC=C2
Melting Point 35°C to 36°C
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 148.23
Physical Form Low Melting Mass
Percent Purity 98%
Chemical Name or Material 5-Methylbenzo[b]thiophene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-256-2

Recommended Storage : Product may darken during storage