Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methylbenzofuroxan, 97%

Click to view available options
Quantity:
1g
5g
19164-41-1
C7H6N2O2
150.137
MFCD00068057
HCWVKDOCXDWFEH-UHFFFAOYSA-N
5-methylbenzofuroxan,benzofurazan, 5-methyl-, 1-oxide,5-methylbenzofurazan-1-oxide,5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate,5-methylbenzofurazan oxide,2,1,3-benzoxadiazole, 5-methyl-, monooxide,5-methylbenzofurazan 1-oxide,acmc-20am21,5-methylbenzfurazan-1-oxide,5-methylbenzofurazan n-oxide
140486
5-methyl-1-oxido-2,1,3-benzoxadiazol-1-ium
CC1=CC2=NO[N+](=C2C=C1)[O-]
19164-41-1
C7H6N2O2
150.137
MFCD00068057
HCWVKDOCXDWFEH-UHFFFAOYSA-N
5-methylbenzofuroxan,benzofurazan, 5-methyl-, 1-oxide,5-methylbenzofurazan-1-oxide,5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate,5-methylbenzofurazan oxide,2,1,3-benzoxadiazole, 5-methyl-, monooxide,5-methylbenzofurazan 1-oxide,acmc-20am21,5-methylbenzfurazan-1-oxide,5-methylbenzofurazan n-oxide
140486
5-methyl-1-oxido-2,1,3-benzoxadiazol-1-ium
CC1=CC2=NO[N+](=C2C=C1)[O-]
Odor Odorless
Quantity 1g
Melting Point 96°C to 98°C
Beilstein 137386
Formula Weight 150.15
Percent Purity 97%
Chemical Name or Material 5-Methylbenzofuroxan

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures