Alfa Aesar™ 5-Methylbenzotriazole

Manufacturer: Alfa Aesar™ 30599-18

Quantity 50g

 View more versions of this product

Catalog No. AA3059918

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 136-85-6
    Formula Weight 133.14
    Chemical Name or Material 5-Methylbenzotriazole

    Specifications

    Formula Weight 133.14
    Health Hazard 2 GHS H Statement
    H311-H302-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several min
    Physical Form Crystalline
    Health Hazard 1 Danger
    Melting Point 72° to 75°C
    Quantity 50g
    TSCA TSCA
    CAS 136-85-6
    Chemical Name or Material 5-Methylbenzotriazole

    GHS H Statement
    H311-H302-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several min

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit