Learn More
5-MethylCytosine Mouse anti-Alligator, Bovine, Human, Mouse, Clone: C.15200006, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259170
Catalog No. NBP259170
Add to cart
Description
Description
5-MethylCytosine Monoclonal antibody specifically detects 5-MethylCytosine in Human, Mouse, Alligator, Bovine samples. It is validated for Methylated DNA Immunoprecipitation, Immunofluorescence.
Specifications
Specifications
5-MethylCytosine
C.15200006
Methylated DNA Immunoprecipitation 0.5-1 ug, Immunofluorescence 1:1000
Supernatant
Mouse
IgG1
100 ug
Store at -80C. Avoid freeze-thaw cycles.
Monoclonal
ChIP, Immunofluorescence
Unconjugated
PBS with 0.05% Sodium Azide
5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl)
The exact immunogen is propietary information.
Size Exclusion Chromatography
RUO
Primary
Human, Mouse, Alligator, Bovine