Learn More
5-MethylCytosine Mouse anti-Drosophila, Human, Mouse, Clone: C.15200003, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259166
Catalog No. NBP259166
Add to cart
Description
Description
5-MethylCytosine Monoclonal antibody specifically detects 5-MethylCytosine in Human, Mouse, Drosophila samples. It is validated for In-situ Hybridization, Dot Blot, Methylated DNA Immunoprecipitation, Immunofluorescence.
Specifications
Specifications
5-MethylCytosine
C.15200003
In-situ Hybridization 1:200 - 1:1000, Dot Blot 1:600, Methylated DNA Immunoprecipitation 1 - 2 ug, Immunofluorescence 1:500
Purified
Mouse
IgG1
50 ug
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
In Situ Hybridization (ISH), Dot Blot, ChIP, Immunofluorescence
Unconjugated
PBS with 0.005% Sodium Azide
5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl)
The exact immunogen is propietary information.
Protein A purified
RUO
Primary
Human, Mouse, Drosophila