Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-MethylCytosine Mouse anti-Human, Mouse, Rat, Zebrafish, Alexa Fluor™ 532, Clone: 4111, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242814AF532

Catalog No. NP242814A32

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

5-MethylCytosine Monoclonal antibody specifically detects 5-MethylCytosine in Human, Mouse, Rat, Zebrafish samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount.
Specifications

Specifications

5-MethylCytosine
4111
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount
Conjugated, Purified
Mouse
IgG1
0.1 mL
Primary
Human, Mouse, Rat, Zebrafish
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl)
The exact sequence is proprietary.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit