Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylfurmethiodide, Tocris Bioscience™

Click to view available options
Quantity:
25mg
This item is not returnable. View return policy
1197-60-0
C9H16INO
281.137
IHTCZSNKQINGDD-UHFFFAOYSA-M
methylfurmetide,methylfurmethide,preparation 7070,5 mft,5-methylfurmethiodide,methylfurmethide iodide,5-methylfurmithide,5-methylfurfuryltrimethylammonium iodide,5-methylfurtrethonium iodide,5-methylfurfuryl trimethylammonium iodide
70973
trimethyl-[(5-methylfuran-2-yl)methyl]azanium;iodide
CC1=CC=C(O1)C[N+](C)(C)C.[I-]
This item is not returnable. View return policy
1197-60-0
C9H16INO
281.137
IHTCZSNKQINGDD-UHFFFAOYSA-M
methylfurmetide,methylfurmethide,preparation 7070,5 mft,5-methylfurmethiodide,methylfurmethide iodide,5-methylfurmithide,5-methylfurfuryltrimethylammonium iodide,5-methylfurtrethonium iodide,5-methylfurfuryl trimethylammonium iodide
70973
trimethyl-[(5-methylfuran-2-yl)methyl]azanium;iodide
CC1=CC=C(O1)C[N+](C)(C)C.[I-]

Potent muscarinic agonist

A potent muscarinic agonist.
Quantity 25mg
Formula Weight 281.14
Percent Purity >99%
Chemical Name or Material 5-Methylfurmethiodide

Recommended Storage : Desiccate at +4°C