Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylindole, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
614-96-0
C9H9N
131.178
MFCD00005680
YPKBCLZFIYBSHK-UHFFFAOYSA-N
5-methylindole,1h-indole, 5-methyl,5-methylindol,indole, 5-methyl,5-methyl indole,unii-kt52363ri5,5-methyl-indole,indole, 2,pubchem7238
11978
5-methyl-1H-indole
CC1=CC2=C(C=C1)NC=C2
614-96-0
C9H9N
131.178
MFCD00005680
YPKBCLZFIYBSHK-UHFFFAOYSA-N
5-methylindole,1h-indole, 5-methyl,5-methylindol,indole, 5-methyl,5-methyl indole,unii-kt52363ri5,5-methyl-indole,indole, 2,pubchem7238
11978
5-methyl-1H-indole
CC1=CC2=C(C=C1)NC=C2
Color White to Beige
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Melting Point 58°C to 61°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, 317
Formula Weight 131.18
Physical Form Crystalline Solid
Percent Purity ≥98.5%
Chemical Name or Material 5-Methylindole, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning