Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylisatin, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
608-05-9
C9H7NO2
161.16
VAJCSPZKMVQIAP-UHFFFAOYSA-N
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
11840
5-methyl-1H-indole-2,3-dione
CC1=CC2=C(C=C1)NC(=O)C2=O
608-05-9
C9H7NO2
161.16
VAJCSPZKMVQIAP-UHFFFAOYSA-N
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
11840
5-methyl-1H-indole-2,3-dione
CC1=CC2=C(C=C1)NC(=O)C2=O
Quantity 5g
Infrared Spectrum Authentic
Melting Point 180.0°C
Assay Percent Range 94% min. (HPLC)
Beilstein 21,509
Solubility Information Solubility in water: insoluble.
Formula Weight 161.16
Percent Purity 95%
Chemical Name or Material 5-Methylisatin

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 210-152-1