Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylnicotinic acid, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
3222-49-9
C7H7NO2
137.14
DJDHHXDFKSLEQY-UHFFFAOYSA-N
5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
256208
5-methylpyridine-3-carboxylic acid
CC1=CN=CC(=C1)C(=O)O
3222-49-9
C7H7NO2
137.14
DJDHHXDFKSLEQY-UHFFFAOYSA-N
5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
256208
5-methylpyridine-3-carboxylic acid
CC1=CN=CC(=C1)C(=O)O
Quantity 1g
Packaging Glass Bottle
Melting Point 211.0°C to 213.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material 5-Methylnicotinic acid

GHS H Statement
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.

GHS Signal Word: Warning

RUO – Research Use Only