Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methylpyridine-2-carbonitrile, ≥97%

Click to view available options
Quantity:
1g
5g
25g
1620-77-5
C7H6N2
118.139
MFCD06200830
LIEQVZZZYLHNRH-UHFFFAOYSA-N
2-cyano-5-methylpyridine,5-methylpicolinonitrile,5-methyl-pyridine-2-carbonitrile,5-methyl-2-pyridinecarbonitrile,6-cyano-3-picoline,2-pyridinecarbonitrile, 5-methyl,2-cyano-5-methylpyridine 2-cyano-5-picoline,5-methypicolinonitrile,pubchem12978,5-methyl picolinonitrile
74183
5-methylpyridine-2-carbonitrile
CC1=CN=C(C=C1)C#N
1620-77-5
C7H6N2
118.139
MFCD06200830
LIEQVZZZYLHNRH-UHFFFAOYSA-N
2-cyano-5-methylpyridine,5-methylpicolinonitrile,5-methyl-pyridine-2-carbonitrile,5-methyl-2-pyridinecarbonitrile,6-cyano-3-picoline,2-pyridinecarbonitrile, 5-methyl,2-cyano-5-methylpyridine 2-cyano-5-picoline,5-methypicolinonitrile,pubchem12978,5-methyl picolinonitrile
74183
5-methylpyridine-2-carbonitrile
CC1=CN=C(C=C1)C#N
Melting Point 72°C to 75°C
Quantity 1g
UN Number UN3439
Beilstein 111125
Formula Weight 118.14
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material 5-Methylpyridine-2-carbonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 216-589-4

TSCA : No

Recommended Storage : Ambient temperatures