Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylpyridine-3-boronic acid, 95%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
173999-18-3
C6H8BNO2
136.95
REONQWGHSQHTAC-UHFFFAOYSA-N
5-methylpyridine-3-boronic acid,5-methyl-3-pyridineboronic acid,5-methylpyridin-3-yl boronic acid,5-methylpyridin-3-yl-3-boronic acid,5-methylpyridine-3-boronicacid,5-methyl-3-pyridyl boronic acid,5-methyl-3-pyridinyl boronic acid,pubchem17082,3-borono-5-methylpyridine,acmc-209e7n
2762708
(5-methylpyridin-3-yl)boronic acid
B(C1=CC(=CN=C1)C)(O)O
173999-18-3
C6H8BNO2
136.95
REONQWGHSQHTAC-UHFFFAOYSA-N
5-methylpyridine-3-boronic acid,5-methyl-3-pyridineboronic acid,5-methylpyridin-3-yl boronic acid,5-methylpyridin-3-yl-3-boronic acid,5-methylpyridine-3-boronicacid,5-methyl-3-pyridyl boronic acid,5-methyl-3-pyridinyl boronic acid,pubchem17082,3-borono-5-methylpyridine,acmc-209e7n
2762708
(5-methylpyridin-3-yl)boronic acid
B(C1=CC(=CN=C1)C)(O)O
Packaging Glass Bottle
Melting Point 300.0°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 136.95
Percent Purity 95%
Chemical Name or Material 5-Methylpyridine-3-boronic acid

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning