Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylquinoxaline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
13708-12-8
C9H8N2
144.177
MFCD00012335
CQLOYHZZZCWHSG-UHFFFAOYSA-N
quinoxaline, 5-methyl,unii-l0ad7t152g,5-methyl quinoxaline,fema no. 3203,8-methylquinoxaline,5-methyl-quinoxaline,acmc-209caq,5-methylquinoxaline,dsstox_cid_27090
61670
5-methylquinoxaline
CC1=C2C(=CC=C1)N=CC=N2
13708-12-8
C9H8N2
144.177
MFCD00012335
CQLOYHZZZCWHSG-UHFFFAOYSA-N
quinoxaline, 5-methyl,unii-l0ad7t152g,5-methyl quinoxaline,fema no. 3203,8-methylquinoxaline,5-methyl-quinoxaline,acmc-209caq,5-methylquinoxaline,dsstox_cid_27090
61670
5-methylquinoxaline
CC1=C2C(=CC=C1)N=CC=N2
Packaging Glass bottle
Density 1.1150g/mL
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 144.18
Chemical Name or Material 5-Methylquinoxaline, 98%

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 237-246-5

TSCA : TSCA