Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylthiazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
3581-89-3
C4H5NS
99.16
RLYUNPNLXMSXAX-UHFFFAOYSA-N
5-methylthiazole,thiazole, 5-methyl,5-methyl-thiazole,5-methyl thiazole,zlchem 1239,acmc-1cjhk,#,ksc222m5d
137980
5-methyl-1,3-thiazole
CC1=CN=CS1
3581-89-3
C4H5NS
99.16
RLYUNPNLXMSXAX-UHFFFAOYSA-N
5-methylthiazole,thiazole, 5-methyl,5-methyl-thiazole,5-methyl thiazole,zlchem 1239,acmc-1cjhk,#,ksc222m5d
137980
5-methyl-1,3-thiazole
CC1=CN=CS1
Boiling Point 141°C to 142°C (993.0 hPa)
Melting Point -40°C
Flash Point 42°C
Color Colorless to Yellow
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Formula Weight 99.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 5-Methylthiazole
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a P

GHS Signal Word: Warning