Alfa Aesar™ 5-Methylthiophene-2-carboxaldehyde, 98+%

Manufacturer: Alfa Aesar™ A1111918

Quantity 50g

 View more versions of this product

Catalog No. AAA1111918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13679-70-4
    Molecular Formula C6H6OS
    Formula Weight 126.18
    MDL Number MFCD00005434
    Beilstein 106896
    Chemical Name or Material 5-Methylthiophene-2-carboxaldehyde

    Specifications

    Formula Weight 126.18
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 13679-70-4
    Boiling Point 92° to 93°C (12mmHg)
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Molecular Formula C6H6OS
    MDL Number MFCD00005434
    Beilstein 106896
    Health Hazard 1 Warning
    Quantity 50g
    Flash Point 87°C (188°F)
    Chemical Name or Material 5-Methylthiophene-2-carboxaldehyde
    Assay ≥98%

    Description

    Applications
    5-Methylthiophene-2-carboxaldehyde is used as a food additive. It is an analytical reagent. The reagent has been synthesized and characterized using IR, 1H NMR and Mass spectral data.

    Solubility
    Soluble in ether and ethanol.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit