Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methylthiophene-2-carboxaldehyde, 98+%

Manufacturer:  Alfa Aesar™ A1111909

Quantity 10g

 View more versions of this product

Catalog No. AAA1111909


  Check Availability
Add to cart

Specifications

Description

Applications
5-Methylthiophene-2-carboxaldehyde is used as a food additive. It is an analytical reagent. The reagent has been synthesized and characterized using IR, 1H NMR and Mass spectral data.

Solubility
Soluble in ether and ethanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

13679-70-4
126.18
106896
C6H6OS
MFCD00005434
5-Methylthiophene-2-carboxaldehyde

Specifications

13679-70-4
126.18
106896
10g
5-Methylthiophene-2-carboxaldehyde
C6H6OS
MFCD00005434
92° to 93°C (12mmHg)
87°C (188°F)
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit