Learn More
BioVision IncSupplier Diversity Partner Link-FAST™ 5 Minutes DNA Ligation Kit: Fast and Convenient Kit for ligation of sticky-end or blunt-end DNA fragments in just 5 minutes at room temperature. No need to prepare buffers or to add ATP or Mg²⁺. 50 reactions. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K90250
The Link-FAST™ 5 minutes DNA Ligation Kit provides a fast and convenient means for ligation of sticky-end or blunt-end DNA fragments in just 5 minutes at room temperature. All necessary reagents required for ligation are provided. There is no need to prepare buffers or to add ATP or Mg²ᶧ. The kit can be used for cloning in plasmid vectors or phage vectors, linker ligation, recirculation of linear DNA, etc. Ligated DNA can be easily analyzed by agarose gel electrophoresis. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9969