Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 5 mm Spinner Turbine

For Bruker™ spectrometers

Manufacturer:  Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 660060056

 View more versions of this product

Catalog No. 03-415-511


Add to cart

Description

Description

  • Matched to Bruker™ turbines
  • Tachometer strip improves life span
  • Temperature range: -20 to 60 °C
Specifications

Specifications

Spinner turbines
Bruker™ spectrometers
Yellow
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit