Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 5 mm Spinner Turbine

For Bruker™ spectrometers

$262.00

Specifications

For Use With (Equipment) Bruker spectrometers
Description Spinner turbines
View More Specs

Products 5
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
03-415-508
View Documents
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™
660060053
Blue
Each for $262.00
Add to Cart
 
03-415-507
View Documents
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™
660060052
Black Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
03-415-509
View Documents
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™
660060054
Green Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
03-415-506
View Documents
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™
660060051
White Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
03-415-511
View Documents
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™
660060056
Yellow Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

  • Matched to Bruker™ turbines
  • Tachometer strip improves life span
  • Temperature range: -20 to 60 °C
Specifications

Specifications

Bruker spectrometers
Spinner turbines
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.