Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[5-(Morpholinomethyl)isoxazol-3-yl]methylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
893748-66-8
C9H15N3O2
197.24
MFCD06804185
JJKIQRMZVVZXSK-UHFFFAOYSA-N
5-morpholinomethyl isoxazol-3-yl methylamine,3-isoxazolemethanamine, 5-4-morpholinylmethyl,5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine,1-5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine
20099025
[5-(morpholin-4-ylmethyl)-1,2-oxazol-3-yl]methanamine
C1COCCN1CC2=CC(=NO2)CN
893748-66-8
C9H15N3O2
197.24
MFCD06804185
JJKIQRMZVVZXSK-UHFFFAOYSA-N
5-morpholinomethyl isoxazol-3-yl methylamine,3-isoxazolemethanamine, 5-4-morpholinylmethyl,5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine,1-5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine
20099025
[5-(morpholin-4-ylmethyl)-1,2-oxazol-3-yl]methanamine
C1COCCN1CC2=CC(=NO2)CN
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 197.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material [5-(Morpholinomethyl)isoxazol-3-yl]methylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger