Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(N,N-Dibenzylglycyl)salicylamide, 98%, Acros Organics

Click to view available options
Quantity:
25g
30566-92-8
C23H22N2O3
374.44
MFCD02181096
DNLPQXRWAKCKPX-UHFFFAOYSA-N
5-2-dibenzylamino acetyl-2-hydroxybenzamide,5-n,n-dibenzylglycyl salicylamide,5-bis benzyl amino acetyl salicylamide,5-2-dibenzylamino acetyl-2-hydroxybenzamide,5-n,n-dibenzylglycyl-2-hydroxybenzamide,5-n,n-dibenzylglcyl salicylamide,5-n,n-dibenzylglycyl salicylamide,5-2-dibenzylamino acetyl salicylamide,5-2-bisbenzylamino acetyl-2-hydroxybenzamide,5-bis phenylmethyl amino acetyl-2-hydroxybenzamide
121727
5-[2-(dibenzylamino)acetyl]-2-hydroxybenzamide
C1=CC=C(C=C1)CN(CC2=CC=CC=C2)CC(=O)C3=CC(=C(C=C3)O)C(=O)N
30566-92-8
C23H22N2O3
374.44
MFCD02181096
DNLPQXRWAKCKPX-UHFFFAOYSA-N
5-2-dibenzylamino acetyl-2-hydroxybenzamide,5-n,n-dibenzylglycyl salicylamide,5-bis benzyl amino acetyl salicylamide,5-2-dibenzylamino acetyl-2-hydroxybenzamide,5-n,n-dibenzylglycyl-2-hydroxybenzamide,5-n,n-dibenzylglcyl salicylamide,5-n,n-dibenzylglycyl salicylamide,5-2-dibenzylamino acetyl salicylamide,5-2-bisbenzylamino acetyl-2-hydroxybenzamide,5-bis phenylmethyl amino acetyl-2-hydroxybenzamide
121727
5-[2-(dibenzylamino)acetyl]-2-hydroxybenzamide
C1=CC=C(C=C1)CN(CC2=CC=CC=C2)CC(=O)C3=CC(=C(C=C3)O)C(=O)N
Quantity 25g
Color Beige
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 374.44
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 5-(N, N-Dibenzylglycyl)salicylamide, 98%