Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitro-1,10-phenanthroline-2,9-dicarboxylic acid hydrate, 98%

Click to view available options
Quantity:
100mg
500mg
164394-23-4
C14H7N3O6
313.225
MFCD00209668
XZQXNUFLUBLEIO-UHFFFAOYSA-N
hydrate,acmc-20am1x,1,10-phenanthroline-2,9-dicarboxylicacid, 5-nitro,5-nitropyridino 3,2-h quinoline-2,9-dicarboxylic acid
7567138
5-nitro-1,10-phenanthroline-2,9-dicarboxylic acid
C1=CC(=NC2=C3C(=C(C=C21)[N+](=O)[O-])C=CC(=N3)C(=O)O)C(=O)O
164394-23-4
C14H7N3O6
313.225
MFCD00209668
XZQXNUFLUBLEIO-UHFFFAOYSA-N
hydrate,acmc-20am1x,1,10-phenanthroline-2,9-dicarboxylicacid, 5-nitro,5-nitropyridino 3,2-h quinoline-2,9-dicarboxylic acid
7567138
5-nitro-1,10-phenanthroline-2,9-dicarboxylic acid
C1=CC(=NC2=C3C(=C(C=C21)[N+](=O)[O-])C=CC(=N3)C(=O)O)C(=O)O
Melting Point ~215°C (decomposition)
Quantity 500mg
Formula Weight 313.23 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 5-Nitro-1,10-phenanthroline-2,9-dicarboxylic acid hydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures