Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitro-2-(3-phenylpropylamino)benzoic acid, 98%

Click to view available options
Quantity:
10mg
50mg
107254-86-4
C16H16N2O4
300.314
MFCD00153851
WBSMIPAMAXNXFS-UHFFFAOYSA-N
nppb,5-nitro-2-3-phenylpropylamino benzoic acid,unii-3a35o9g3yz,5-nitro-2-3-phenylpropyl amino benzoic acid,benzoic acid,5-nitro-2-3-phenylpropyl amino,benzoic acid, 5-nitro-2-3-phenylpropyl amino,5-nitro-2-3-phenylpropylamino-benzoic acid,5-nitro-2-phenylpropylaminobenzoic acid nppb,5-nitro-2-3-phenylpropylamino benzoic acid nppb,spectrum_001814
4549
CHEBI:34457
5-nitro-2-(3-phenylpropylamino)benzoic acid
C1=CC=C(C=C1)CCCNC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)O
107254-86-4
C16H16N2O4
300.314
MFCD00153851
WBSMIPAMAXNXFS-UHFFFAOYSA-N
nppb,5-nitro-2-3-phenylpropylamino benzoic acid,unii-3a35o9g3yz,5-nitro-2-3-phenylpropyl amino benzoic acid,benzoic acid,5-nitro-2-3-phenylpropyl amino,benzoic acid, 5-nitro-2-3-phenylpropyl amino,5-nitro-2-3-phenylpropylamino-benzoic acid,5-nitro-2-phenylpropylaminobenzoic acid nppb,5-nitro-2-3-phenylpropylamino benzoic acid nppb,spectrum_001814
4549
CHEBI:34457
5-nitro-2-(3-phenylpropylamino)benzoic acid
C1=CC=C(C=C1)CCCNC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)O
Color Yellow
Quantity 10mg
Melting Point 178°C to 180°C
Merck Index 14,6725
Formula Weight 300.31
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Nitro-2-(3-phenylpropylamino)benzoic acid