Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitro-2-furaldehyde, 99%

Click to view available options
Quantity:
25g
50g
698-63-5
C5H3NO4
141.082
MFCD00003230
SXINBFXPADXIEY-UHFFFAOYSA-N
5-nitro-2-furaldehyde,5-nitrofurfural,nitrofurfural,2-furancarboxaldehyde, 5-nitro,5-nitrofurfuraldehyde,5-nitro-2-furfural,2-furaldehyde, 5-nitro,5-nitrofuraldehyde,5-nitro-2-furancarboxaldehyde,furfural, 5-nitro
12774
CHEBI:34459
5-nitrofuran-2-carbaldehyde
C1=C(OC(=C1)[N+](=O)[O-])C=O
698-63-5
C5H3NO4
141.082
MFCD00003230
SXINBFXPADXIEY-UHFFFAOYSA-N
5-nitro-2-furaldehyde,5-nitrofurfural,nitrofurfural,2-furancarboxaldehyde, 5-nitro,5-nitrofurfuraldehyde,5-nitro-2-furfural,2-furaldehyde, 5-nitro,5-nitrofuraldehyde,5-nitro-2-furancarboxaldehyde,furfural, 5-nitro
12774
CHEBI:34459
5-nitrofuran-2-carbaldehyde
C1=C(OC(=C1)[N+](=O)[O-])C=O
Applications
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
Sensitivity Air and light sensitive
Refractive Index 1.59
Density 1.349
Flash Point 33°C (91°F)
Melting Point 34°C to 38°C
Boiling Point 228°C
Quantity 25g
Beilstein 120539
Solubility Information Sparingly soluble in water.
Formula Weight 141.09
Percent Purity 99%
Chemical Name or Material 5-Nitro-2-furaldehyde
Show More Show Less

GHS H Statement
H341
Suspected of causing genetic defects.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 211-816-3

RTECSNumber : LT7200000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon