Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitro-2-furaldehyde diacetate, 98+%

Click to view available options
Quantity:
100g
500g
2500g
92-55-7
C9H9NO7
243.171
MFCD00003244
HSXKWKJCZNRMJO-UHFFFAOYSA-N
5-nitro-2-furaldehyde diacetate,2-diacetoxymethyl-5-nitrofuran,5-nitro-2-furanmethanediol diacetate,5-nitrofurfural diacetate,5-nitrofurfurylidene diacetate,5-nitrofuran-2-yl methylene diacetate,nitrofuraldehyde diacetate,5-nitro-2-furfural diacetate,5-nitrofuraldehyde diacetate,5-nitro-2-furfuraldehyde diacetate
7097
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C
92-55-7
C9H9NO7
243.171
MFCD00003244
HSXKWKJCZNRMJO-UHFFFAOYSA-N
5-nitro-2-furaldehyde diacetate,2-diacetoxymethyl-5-nitrofuran,5-nitro-2-furanmethanediol diacetate,5-nitrofurfural diacetate,5-nitrofurfurylidene diacetate,5-nitrofuran-2-yl methylene diacetate,nitrofuraldehyde diacetate,5-nitro-2-furfural diacetate,5-nitrofuraldehyde diacetate,5-nitro-2-furfuraldehyde diacetate
7097
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C
Sensitivity Light sensitive
Melting Point 87°C to 92°C
Quantity 2500g
Beilstein 255089
Formula Weight 243.17
Percent Purity ≥98%
Chemical Name or Material 5-Nitro-2-furaldehyde diacetate

GHS H Statement
H341
Suspected of causing genetic defects.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

EINECSNumber : 202-166-1

RTECSNumber : LU1940000

TSCA : Yes

Recommended Storage : Ambient temperatures