Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitro-2-furaldehyde diacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic bottle
92-55-7
C9H9NO7
243.17
MFCD00003244
HSXKWKJCZNRMJO-UHFFFAOYSA-N
5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate
7097
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C
92-55-7
C9H9NO7
243.17
MFCD00003244
HSXKWKJCZNRMJO-UHFFFAOYSA-N
5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate
7097
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100g
Melting Point 89.0°C to 93.0°C
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 243.17
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Nitro-2-furaldehyde diacetate, 98%
Show More Show Less

EINECSNumber : 202-166-1

RTECSNumber : LU1940000

TSCA : TSCA