Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Nitro-2-furaldehyde semicarbazone, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
250 g
Description
Bactericidal compound used as an antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼240°C (decomposition) |
Quantity | 25 g |
Beilstein | 86403 |
Sensitivity | Light sensitive |
Merck Index | 14,6600 |
Formula Weight | 198.14 |
Percent Purity | ≥98% |
Chemical Name or Material | 5-Nitro-2-furaldehyde semicarbazone |
Safety and Handling
Hazard Category | H302-H317 |
Hazard Statement | GHS H Statement H301-H341-H351-H361 Toxic if swallowed. Suspected of causing genetic defects. Suspected of causing cancer. Suspected of damaging fertility or the unborn child. |
Precautionary Statement | P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c |
EINECSNumber | 200-443-1 |
RTECSNumber | LT7700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction