Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitro-2-furaldoxime, 97%

Click to view available options
Quantity:
10g
50g
555-15-7
C5H4N2O4
156.097
MFCD00198004
PTBKFATYSVLSSD-ZZXKWVIFSA-N
nifuroxime,5-nitro-2-furaldehyde oxime,usaf ea-5,2-furaldehyde, 5-nitro-, oxime,2-furaldoxime, 5-nitro,5-nitro-2-furaldoxime,carbazylquinone,carbilquinone,5-nitrofuran-2-carbaldehyde oxime,5-nitro-2-furancarboxaldehyde oxime
6478035
(NE)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
C1=C(OC(=C1)[N+](=O)[O-])C=NO
555-15-7
C5H4N2O4
156.097
MFCD00198004
PTBKFATYSVLSSD-ZZXKWVIFSA-N
nifuroxime,5-nitro-2-furaldehyde oxime,usaf ea-5,2-furaldehyde, 5-nitro-, oxime,2-furaldoxime, 5-nitro,5-nitro-2-furaldoxime,carbazylquinone,carbilquinone,5-nitrofuran-2-carbaldehyde oxime,5-nitro-2-furancarboxaldehyde oxime
6478035
(NE)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
C1=C(OC(=C1)[N+](=O)[O-])C=NO
Sensitivity Light sensitive
Quantity 10g
Beilstein 83037
Merck Index 14,6536
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material 5-Nitro-2-furaldoxime

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 209-082-4

RTECSNumber : LT7525000

TSCA : No

Recommended Storage : Ambient temperatures