Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 5-Nitro-2-furonitrile, 97%
Click to view available options
Quantity:
5g
25g
Specifications
Melting Point | 61°C to 64°C |
Quantity | 5g |
UN Number | UN3439 |
Beilstein | 123933 |
Formula Weight | 138.08 |
Percent Purity | 97% |
Chemical Name or Material | 5-Nitro-2-furonitrile |
Safety and Handling
GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P361-P363-P501c
H302+H332-H311
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures