Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitrobarbituric acid trihydrate, 95%

Click to view available options
Quantity:
5g
25g
6209-44-5
C4H9N3O8
227.129
MFCD00149458
MXFRQEURRMBYRS-UHFFFAOYSA-N
5-nitrobarbituric acid trihydrate,5-nitro-1,3-diazinane-2,4,6-trione trihydrate,pubchem21911,acmc-1ba25,5-nitropyrimidine-2,4,6 1h,3h,5h-trione trihydrate
60145881
5-nitro-1,3-diazinane-2,4,6-trione;trihydrate
C1(C(=O)NC(=O)NC1=O)[N+](=O)[O-].O.O.O
6209-44-5
C4H9N3O8
227.129
MFCD00149458
MXFRQEURRMBYRS-UHFFFAOYSA-N
5-nitrobarbituric acid trihydrate,5-nitro-1,3-diazinane-2,4,6-trione trihydrate,pubchem21911,acmc-1ba25,5-nitropyrimidine-2,4,6 1h,3h,5h-trione trihydrate
60145881
5-nitro-1,3-diazinane-2,4,6-trione;trihydrate
C1(C(=O)NC(=O)NC1=O)[N+](=O)[O-].O.O.O
Quantity 25g
Melting Point 182°C to 184°C (decomposition)
Merck Index 14,6586
Formula Weight 227.14 (173.08 Anhydrous)
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 5-Nitrobarbituric acid trihydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA