Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitroindoline, 97%

Click to view available options
Quantity:
5g
25g
32692-19-6
C8H8N2O2
164.164
MFCD00005709
WJQWYAJTPPYORB-UHFFFAOYSA-N
5-nitroindoline,1h-indole, 2,3-dihydro-5-nitro,indoline, 5-nitro,2,3-dihydro-5-nitro-1h-indole,ccris 3306,5-nitro-2,3-dihydro-indole,5-nitroindolin,5-nitro indoline,pubchem7442,acmc-1aera
36219
5-nitro-2,3-dihydro-1H-indole
C1CNC2=C1C=C(C=C2)[N+](=O)[O-]
32692-19-6
C8H8N2O2
164.164
MFCD00005709
WJQWYAJTPPYORB-UHFFFAOYSA-N
5-nitroindoline,1h-indole, 2,3-dihydro-5-nitro,indoline, 5-nitro,2,3-dihydro-5-nitro-1h-indole,ccris 3306,5-nitro-2,3-dihydro-indole,5-nitroindolin,5-nitro indoline,pubchem7442,acmc-1aera
36219
5-nitro-2,3-dihydro-1H-indole
C1CNC2=C1C=C(C=C2)[N+](=O)[O-]
Melting Point 92°C to 94°C
Quantity 5g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 5-Nitroindoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning