Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroisatin 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
611-09-6
C8H4N2O4
192.13
MFCD00005720
UNMYHYODJHKLOC-UHFFFAOYSA-N
5-nitroisatin,5-nitroindoline-2,3-dione,5-nitroindole-2,3-dione,1h-indole-2,3-dione, 5-nitro,indole-2,3-dione, 5-nitro,isatin, 5-nitro,n-nitroisatin,ccris 4031,2,3-dihydro-5-nitroindole-2,3-dione,5-nitro-2,3-dihydro-1h-indole-2,3-dione
4669250
5-nitro-1H-indole-2,3-dione
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2
611-09-6
C8H4N2O4
192.13
MFCD00005720
UNMYHYODJHKLOC-UHFFFAOYSA-N
5-nitroisatin,5-nitroindoline-2,3-dione,5-nitroindole-2,3-dione,1h-indole-2,3-dione, 5-nitro,indole-2,3-dione, 5-nitro,isatin, 5-nitro,n-nitroisatin,ccris 4031,2,3-dihydro-5-nitroindole-2,3-dione,5-nitro-2,3-dihydro-1h-indole-2,3-dione
4669250
5-nitro-1H-indole-2,3-dione
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2
Melting Point 251°C
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange to Orange-Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 21, 456
Formula Weight 192.13
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 5-Nitroisatin, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-252-5

RTECSNumber : NL7970000