Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitroisoquinoline, 98+%

Click to view available options
Quantity:
5g
25g
607-32-9
C9H6N2O2
174.159
MFCD00006905
PYGMPFQCCWBTJQ-UHFFFAOYSA-N
isoquinoline, 5-nitro,5-nitro-isoquinoline,5-nitro isoquinoline,5-nitroisochinolin,pubchem6272,maybridge1_001820,acmc-1av1q,5-nitroisoquinoline,pygmpfqccwbtjq-uhfffaoysa
69085
5-nitroisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)[N+](=O)[O-]
607-32-9
C9H6N2O2
174.159
MFCD00006905
PYGMPFQCCWBTJQ-UHFFFAOYSA-N
isoquinoline, 5-nitro,5-nitro-isoquinoline,5-nitro isoquinoline,5-nitroisochinolin,pubchem6272,maybridge1_001820,acmc-1av1q,5-nitroisoquinoline,pygmpfqccwbtjq-uhfffaoysa
69085
5-nitroisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)[N+](=O)[O-]
Quantity 5g
Melting Point 106°C to 110°C
Beilstein 134858
Formula Weight 174.16
Percent Purity ≥98%
Chemical Name or Material 5-Nitroisoquinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H500

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-133-8

TSCA : No

Recommended Storage : Ambient temperatures