Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroisoquinoline, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
607-32-9
C9H6N2O2
174.159
MFCD00006905
PYGMPFQCCWBTJQ-UHFFFAOYSA-N
isoquinoline, 5-nitro,5-nitro-isoquinoline,5-nitro isoquinoline,5-nitroisochinolin,pubchem6272,maybridge1_001820,acmc-1av1q,5-nitroisoquinoline,pygmpfqccwbtjq-uhfffaoysa
69085
5-nitroisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)[N+](=O)[O-]
607-32-9
C9H6N2O2
174.159
MFCD00006905
PYGMPFQCCWBTJQ-UHFFFAOYSA-N
isoquinoline, 5-nitro,5-nitro-isoquinoline,5-nitro isoquinoline,5-nitroisochinolin,pubchem6272,maybridge1_001820,acmc-1av1q,5-nitroisoquinoline,pygmpfqccwbtjq-uhfffaoysa
69085
5-nitroisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)[N+](=O)[O-]
Color Yellow
Packaging Glass bottle
Melting Point 106°C to 110°C
Infrared Spectrum Authentic
Quantity 10g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 20, 386
Formula Weight 174.16
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 5-Nitroisoquinoline, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-133-8