Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroquinoline 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
607-34-1
C9H6N2O2
174.159
MFCD00006790
NDDZXHOCOKCNBM-UHFFFAOYSA-N
quinoline, 5-nitro,5-nitro-quinoline,zlchem 399,pubchem5821,acmc-209mlr,5-nitroquinoline,5-20-07-00325 beilstein handbook reference,ksc355k8h,7ze
11829
5-nitroquinoline
C1=CC2=C(C=CC=N2)C(=C1)[N+](=O)[O-]
607-34-1
C9H6N2O2
174.159
MFCD00006790
NDDZXHOCOKCNBM-UHFFFAOYSA-N
quinoline, 5-nitro,5-nitro-quinoline,zlchem 399,pubchem5821,acmc-209mlr,5-nitroquinoline,5-20-07-00325 beilstein handbook reference,ksc355k8h,7ze
11829
5-nitroquinoline
C1=CC2=C(C=CC=N2)C(=C1)[N+](=O)[O-]
Quantity 25g
Packaging Glass bottle
Melting Point 70°C to 73°C
Color Yellow
Infrared Spectrum Authentic
Sulfated Ash 0.1% max.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, 371
Formula Weight 174.16
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-Nitroquinoline, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Suspected of causing genetic defects.
Causes skin irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-134-3

RTECSNumber : VC1850000